missing translation for 'onlineSavingsMsg'
Learn More

Papaverine Hydrochloride, Mikromol™

Product Code. 18032940
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18032940 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18032940 Supplier Mikromol Supplier No. MM1225.00
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Papaverine Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Papaverine Hydrochloride
CAS 61-25-6
Analyte or Component Names Papaverine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C20 H21 N O4 . Cl H
Certifications/Compliance ISO 17034
Synonym Papaverine hydrochloride, 1-(3,4-Dimethoxybenzyl)-6,7-dimethoxyisoquinoline hydrochloride, Isoquinoline, 1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-, hydrochloride, Isoquinoline, 6,7-dimethoxy-1-veratryl-, hydrochloride, 1-(3,4-Dimethoxybenzyl)-6,7-dimethoxyisoquinoline hydrochloride, 6,7,3',4'-Tetramethoxy-1-benzylisoquinoline hydrochloride, 6,7-Dimethoxy-1-(3,4-dimethoxybenzyl)isoquinoline hydrochloride, 6,7-Dimethoxy-1-veratrylisoquinoline hydrochloride, Artegodan, Cardiospan, Cardoverina, Cepaverin, Cerebid, Cerespan, Dispamil, Drapavel, Dynovas, Myobid, Optenyl, Pameion, Panergon, Papavarine chlorhydrate, Papaverinium chloride, Papaversan, Paptial TR, Pavabid, Pavacap, Pavacen, Pavadel, Pavagen, Pavakey, Pavased, Paveron, Ro-Papav, Spasmo Nit, Therapav, Vasal, Vasorin, Vasospan, Noscapine Hydrochloride Hydrate Impurity A as Hydrochloride, Noscapine Impurity A as Hydrochloride, Noscapine Impurity A as Hydrochloride
InChI Formula InChI=1S/C20H21NO4.ClH/c1-22-17-6-5-13(10-18(17)23-2)9-16-15-12-20(25-4)19(24-3)11-14(15)7-8-21-16;/h5-8,10-12H,9H2,1-4H3;1H
SMILES Cl.COc1ccc(Cc2nccc3cc(OC)c(OC)cc23)cc1OC
IUPAC Name 1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxyisoquinoline;hydrochloride
Molecular Weight (g/mol) 375.85
Quantity 250 mg
Formula Weight 375.1237 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.