missing translation for 'onlineSavingsMsg'
Learn More

Paroxetine EP Impurity B, Mikromol™

Product Code. 30458482
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30458482 4 x 25 mg 100mg
17930596 25 mg 25mg
17922406 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30458482 Supplier Mikromol Supplier No. MM1032.0200254AMP
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Paroxetine EP Impurity B, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sesamol
CAS 533-31-3
Analyte or Component Names Sesamol
Shipping Condition Room Temperature
Molecular Formula C7 H6 O3
Certifications/Compliance ISO 17025
Synonym 1,3-Benzodioxol-5-ol (Sesamol), Paroxetine Hydrochloride Anhydrous Imp. B (EP), Paroxetine Imp. B (EP), 1,3-Benzodioxol-5-ol, Sesamol, BRL-43782, Paroxetine Related, 3,4-Methylenedioxyphenol, Sesamol (BRL43782), BP Paroxetine Impurity Standard, Paroxetine Hydrochloride Impurity B, Paroxetine Impurity B
InChI Formula InChI=1S/C7H6O3/c8-5-1-2-6-7(3-5)10-4-9-6/h1-3,8H,4H2
Solution Type Neat
SMILES Oc1ccc2OCOc2c1
IUPAC Name 1,3-benzodioxol-5-ol
Molecular Weight (g/mol) 138.12
Quantity 4 x 25 mg
Formula Weight 138.0317 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.