missing translation for 'onlineSavingsMsg'
Learn More

Paroxetine Hydrochloride Hemihydrate 1.0 mg/ml in Methanol (as anhydrous free base), LoGiCal

Product Code. 30458527
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30458527 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458527 Supplier LoGiCal Supplier No. LGCAMP0264.0001
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Paroxetine Hydrochloride Hemihydrate 1.0 mg/ml in Methanol (as anhydrous free base)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Paroxetine Hydrochloride Hemihydrate
CAS 110429-35-1
Analyte or Component Names Paroxetine Hydrochloride Hemihydrate
Shipping Condition Room Temperature
Molecular Formula 2 C19 H20 F N O3 . 2 Cl H . H2 O
Certifications/Compliance ISO 17034
Synonym Paroxetine Hydrochloride Hemihydrate, Piperidine, 3-[(1,3-benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)-, hydrochloride, hydrate (2:2:1), (3 S,4 R)-, Piperidine, 3-[(1,3-benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)-, hydrochloride, hydrate (2:1), (3 S,4 R)- (9 CI), Piperidine, 3-[(1,3-benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)-, hydrochloride, hydrate (2:1), (3 S-trans)-, (-)-trans-4-(4-Fluorophenyl)-3-(3,4-methylenedioxyphenoxymethyl)piperidine hydrochloride hemihydrate, BRL-29060 A, BRL-29060 A, BRL-29060 A, BRL-29060 A, BRL-29060 A, BRL-29060 A, BRL-29060 A
InChI Formula InChI=1 S/2C19H20FNO3.2 ClH.H2O/c2*20-15-3-1-13(2-4-15)17-7-8-21-10-14(17)11-22-16-5-6-18-19(9-16)24-12-23-18;;;/h2*1-6,9,14,17,21 H,7-8,10-12H2;2*1 H;1H2/t2*14-,17-;;;/m00.../s1
SMILES O.Cl.Cl.Fc1ccc(cc1)[C@@H]2 CCNC[C@H]2 COc3ccc4OCOc4c3.Fc5ccc(cc5)[C@@H]6 CCNC[C@H]6 COc7ccc8OCOc8c7
IUPAC Name (3 S,4 R)-3-(1,3-benzodioxol-5-yloxymethyl)-4-(4-fluorophenyl)piperidine;hydrate;dihydrochloride
Solvent or Matrix Methanol
Industry Type Forensics
Molecular Weight (g/mol) 749.67
Quantity 1 mL
Formula Weight 748.2494 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.