missing translation for 'onlineSavingsMsg'
Learn More

Patched 2/PTCH2 Antibody (OTI8B5), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18161587 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18161587 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18161587 Supplier Novus Biologicals Supplier No. NBP273255AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Patched 2/PTCH2 Monoclonal specifically detects Patched 2/PTCH2 in Human, Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Patched 2/PTCH2
Applications Western Blot
Classification Monoclonal
Clone OTI8B5
Conjugate Alexa Fluor 750
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias patched (Drosophila) homolog 2, patched 2, patched homolog 2, patched homolog 2 (Drosophila), protein patched homolog 2, PTC2
Gene Symbols PTCH2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 79-392 of human Patched 2/PTCH2 (NP_003729) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 8643.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.