missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PAX1 Polyclonal antibody specifically detects PAX1 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | PAX1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | paired box 1, paired domain gene HuP48 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 300-400 of human PAX1 (NP_006183.2). GTAYSPKMEDWAGVNR |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?