Learn More
Invitrogen™ Paxillin Recombinant Rabbit Monoclonal Antibody (23GB2690), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA553295
 View more versions of this product
Product Code. 30089968
  • 207.00 € / 20µL
Description
Paxillin is a focal adhesion protein and a substrate for several tyrosine kinases such as src, FAK, and p210BRC/ABL. The tyrosine phosphorylation of paxillin is affected by conditions that change cell-cell adhesion. Paxillin associates tightly with FAK and Crk through its SH2 domain. This interaction is independent of the extracellular matrix. Although paxillin was initially discovered in fibroblasts, its phosphorylation may also be important during neurite extension during differentiation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Paxillin
Recombinant Monoclonal
0.12 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P49023, Q66H76, Q8VI36
PXN
A synthesized peptide derived from human Paxillin (1-39AA).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
23GB2690
Unconjugated
PXN
adaptor protein; AW108311; AW123232; FLJ16691; hCG_1778014; I79_001229; myocardial ischemic preconditioning associated protein 7; Pax; PAX-1; PAXI; Paxillin; paxillin a; paxillin alpha; pxn; pxna; testicular tissue protein Li 134; TGFB1I1; transforming growth factor beta 1 induced transcript 1; wu:fw71f12
Rabbit
Affinity chromatography
RUO
19303, 360820, 5829
-20°C