missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Paxillin Recombinant Rabbit Monoclonal Antibody (23GB2690), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
207.00 € - 562.00 €
Specifications
| Antigen | Paxillin |
|---|---|
| Clone | 23GB2690 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089918
![]() |
Invitrogen™
MA553294 |
100 μL |
562.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089968
|
Invitrogen™
MA553295 |
20 μL |
207.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Paxillin Recombinant Monoclonal antibody specifically detects Paxillin in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| Paxillin | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P49023, Q66H76, Q8VI36 | |
| 19303, 360820, 5829 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB2690 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| adaptor protein; AW108311; AW123232; FLJ16691; hCG_1778014; I79_001229; myocardial ischemic preconditioning associated protein 7; Pax; PAX-1; PAXI; Paxillin; paxillin a; paxillin alpha; pxn; pxna; testicular tissue protein Li 134; TGFB1I1; transforming growth factor beta 1 induced transcript 1; wu:fw71f12 | |
| A synthesized peptide derived from human Paxillin (1-39AA). | |
| Primary | |
| -20°C | |
| PXN |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title