missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PBR Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-45769-0.025MG
This item is not returnable.
View return policy
Description
PBR Polyclonal specifically detects PBR in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PBR | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.2), 0.5% BSA, 30% Glycerol with 0.01% Thimerosal | |
| benzodiazepine peripheral binding site, BZRPDBI, IBP, MBRPBS, mDRC, Mitochondrial benzodiazepine receptor, PBRBPBS, PBRS, peripheral benzodiazepine receptor, Peripheral-type benzodiazepine receptor, pk18, PKBSbenzodiazapine receptor (peripheral), PTBR, translocator protein, translocator protein (18kDa) | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 706 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry | |
| 0.5 mg/ml | |
| Western Blot, Immunohistochemistry | |
| BAA04749 | |
| TSPO | |
| Synthetic peptide surrounding amino acid 81 of mouse PBR (BAA04749). | |
| 0.025 mg | |
| Primary | |
| Mouse, Rat, Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction