missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PBR Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
236.00 € - 558.00 €
Specifications
| Antigen | PBR |
|---|---|
| Concentration | 0.5 mg/ml |
| Dilution | Western Blot, Immunohistochemistry |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18022816
|
Novus Biologicals
NBP1-45769-0.025MG |
0.025 mg |
236.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18748793
|
Novus Biologicals
NBP1-45769 |
0.1 mg |
558.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PBR Polyclonal specifically detects PBR in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| PBR | |
| Western Blot, Immunohistochemistry | |
| Polyclonal | |
| Rabbit | |
| Mouse, Rat, Human | |
| BAA04749 | |
| 706 | |
| Synthetic peptide surrounding amino acid 81 of mouse PBR (BAA04749). | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| 0.5 mg/ml | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2), 0.5% BSA, 30% Glycerol with 0.01% Thimerosal | |
| benzodiazepine peripheral binding site, BZRPDBI, IBP, MBRPBS, mDRC, Mitochondrial benzodiazepine receptor, PBRBPBS, PBRS, peripheral benzodiazepine receptor, Peripheral-type benzodiazepine receptor, pk18, PKBSbenzodiazapine receptor (peripheral), PTBR, translocator protein, translocator protein (18kDa) | |
| TSPO | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title