missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCCB Mouse anti-Pig, Rat, Mouse, Human, CoraLite 488, Clone: 1A9A5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66501-100UL
This item is not returnable.
View return policy
Description
PCCB Monoclonal antibody apecifically detetects PCCB antigen in Pig, Rat, Mouse, Human Samples. It is validated for Immunofluorescence.Specifications
| PCCB | |
| Monoclonal | |
| 2000 μg/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| PCCB | |
| PCCB fusion protein Ag1608 | |
| 100 μL | |
| 5096 | |
| Store at -20°C. Avoid exposure to light. | |
| IgG1 |
| Immunofluorescence | |
| 1A9A5 | |
| CoraLite 488 | |
| BC013768 | |
| Mouse | |
| Protein G purification | |
| Primary | |
| Pig, Rat, Mouse, Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction