missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCCB Mouse anti-Pig, Rat, Mouse, Human, CoraLite 488, Clone: 1A9A5, Proteintech
Mouse Monoclonal Antibody
145.00 € - 450.00 €
Specifications
| Antigen | PCCB |
|---|---|
| Clone | 1A9A5 |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | CoraLite 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17781289
|
Proteintech
CL488-66501-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17639866
|
Proteintech
CL488-66501-100UL |
100 μL |
450.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PCCB Monoclonal antibody apecifically detetects PCCB antigen in Pig, Rat, Mouse, Human Samples. It is validated for Immunofluorescence.Specifications
| PCCB | |
| Immunofluorescence | |
| CoraLite 488 | |
| Mouse | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| 5096 | |
| PCCB fusion protein Ag1608 | |
| Primary | |
| Store at -20°C. Avoid exposure to light. |
| 1A9A5 | |
| Monoclonal | |
| Liquid | |
| Pig, Rat, Mouse, Human | |
| BC013768 | |
| PCCB | |
| IgG1 | |
| Protein G purification |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title