missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PCM1 Monoclonal antibody specifically detects PCM1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | PCM1 |
| Applications | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | CL0206 |
| Conjugate | DyLight 650 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | hPCM-1, pericentriolar material 1, pericentriolar material 1 protein, PTC4PCM-1 |
| Host Species | Mouse |
| Immunogen | This antibody was developed using a recombinant protein derived from Q15154, with the exact immunogen sequence remaining proprietary. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?