missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCNA Antibody (rPCNA/8859), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23682-100ug
This item is not returnable.
View return policy
Description
PCNA Monoclonal antibody specifically detects PCNA in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| PCNA | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| cyclin, DNA polymerase delta auxiliary protein, MGC8367, proliferating cell nuclear antigen | |
| Recombinant full-length human PCNA protein | |
| 100 μg | |
| Autophagy, Base Excision Repair, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Polymerases, DNA Repair, Loading Controls, Phospho Specific, Stem Cell Markers | |
| 5111 | |
| Store at 4°C. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rPCNA/8859 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction