missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCNA Antibody (rPCNA/8859), Novus Biologicals™
Mouse Monoclonal Antibody
232.00 € - 462.00 €
Specifications
| Antigen | PCNA |
|---|---|
| Clone | rPCNA/8859 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18602435
|
Novus Biologicals
NBP3-23682-20ug |
20 μg |
232.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18686704
|
Novus Biologicals
NBP3-23682-100ug |
100 μg |
462.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PCNA Monoclonal antibody specifically detects PCNA in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| PCNA | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Autophagy, Base Excision Repair, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Polymerases, DNA Repair, Loading Controls, Phospho Specific, Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| 5111 | |
| IgG1 κ | |
| Protein A or G purified |
| rPCNA/8859 | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| cyclin, DNA polymerase delta auxiliary protein, MGC8367, proliferating cell nuclear antigen | |
| Recombinant full-length human PCNA protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title