missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PDRG1 Monoclonal antibody specifically detects PDRG1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | PDRG1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | OTI4F12 |
| Conjugate | mFluor Violet 610 SE |
| Dilution | Flow Cytometry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | C20orf126p53 and DNA damage regulated 1, chromosome 20 open reading frame 126, dJ310O13.3, p53 and DNA-damage regulated 1, PDRGp53 and DNA damage-regulated protein 1 |
| Host Species | Mouse |
| Immunogen | Full length human recombinant protein of human PDRG1(NP_110442) produced in HEK293T cell. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?