missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentifylline, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Pentifylline |
| CAS | 1028-33-7 |
| Analyte or Component Names | Pentifylline |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H20 N4 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1H-Purine-2,6-dione, 1-hexyl-3,7-dihydro-3,7-dimethyl-, Theobromine, 1-hexyl- (6CI,7CI,8CI), 1-Hexyl-3,7-dimethylxanthine, 1-Hexyltheobromine, 1-n-Hexyl-3,7-dimethyl-xanthine, 3,7-Dimethyl-1-hexyl-1H,3H-purin-2,6-dione, Cosadon, Cosaldon, Hexyltheobromine, Pentifylline, SK 7, SK 7 (pharmaceutical) |
| InChI Formula | InChI=1S/C13H20N4O2/c1-4-5-6-7-8-17-12(18)10-11(14-9-15(10)2)16(3)13(17)19/h9H,4-8H2,1-3H3 |
| Solution Type | Neat |
| SMILES | CCCCCCN1C(=O)N(C)c2ncn(C)c2C1=O |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction