missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEO1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93513-0.02ml
This item is not returnable.
View return policy
Description
PEO1 Polyclonal antibody specifically detects PEO1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PEO1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| ATXN8, chromosome 10 open reading frame 2, EC 3.6.4.12, FLJ21832, infantile onset spinocerebellar ataxia (autosomal recessive), IOSCA, mitochondrial, MTDPS7, PEO1PEOA3, TWINKLE | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 135-335 of human PEO1 (NP_068602.2). GAREGFLLSKAPEFEDSEEVRRIWNRAIPLWELPDQEEVQLADTMFGLTKVTDDTLKRFSVRYLRPARSLVFPWFSPGGSGLRGLKLLEAKCQGDGVSYEETTIPRPSAYHNLFGLPLISRRDAEVVLTSRELDSLALNQSTGLPTLTLPRGTTCLPPALLPYLEQFRRIVFWLGDDLRSWEAAKLFARKLNPKRCFLVRP | |
| 0.02 mL | |
| Endocrinology, Neurodegeneration, Neuroscience | |
| 56652 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction