missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peptidylglycine alpha-Amidating Monooxygenase/PAM Antibody, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Peptidylglycine alpha-Amidating Monooxygenase/PAM Polyclonal specifically detects Peptidylglycine alpha-Amidating Monooxygenase/PAM in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | Peptidylglycine alpha-Amidating Monooxygenase/PAM |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Accession No. | P19021 |
| Gene Alias | PAL, pancreatic peptidylglycine alpha-amidating monooxygenase, peptidyl alpha-amidating enzyme, peptidyl-alpha-hydroxyglycine alpha-amidating lyase, peptidylamidoglycolate lyase, peptidylglycine 2-hydroxylase, peptidyl-glycine alpha-amidating monooxygenase, peptidylglycine alpha-amidating monooxygenase, peptidylglycine alpha-hydroxylating monooxygenase, PHM |
| Gene Symbols | PAM |
| Host Species | Rabbit |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?