missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perazine Dimaleate 1.0 mg/ml in Dimethyl Sulfoxide (as free base), LoGiCal
Specifications
Specifications
| Chemical Name or Material | Perazine Dimaleate |
| CAS | 14516-56-4 |
| Analyte or Component Names | Perazine Dimaleate |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C20 H25 N3 S . 2 C4 H4 O4 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Perazine Dimaleate, 10H-Phenothiazine, 10-[3-(4-methyl-1-piperazinyl)propyl]-, (Z)-2-butenedioate (1:2), Phenothiazine, 10-[3-(4-methyl-1-piperazinyl)propyl]-, dimaleate (6CI), Phenothiazine, 10-[3-(4-methyl-1-piperazinyl)propyl]-, maleate (1:2) (8CI), Perazine dimaleate, Perazine maleate, Pernazinum, Prochlorperazine Maleate Impurity B as Dimaleate, Prochlorperazine Impurity B as Dimaleate |
| InChI Formula | InChI=1S/C20H25N3S.2C4H4O4/c1-21-13-15-22(16-14-21)11-6-12-23-17-7-2-4-9-19(17)24-20-10-5-3-8-18(20)23;2*5-3(6)1-2-4(7)8/h2-5,7-10H,6,11-16H2,1H3;2*1-2H,(H,5,6)(H,7,8)/b;2*2-1- |
| Solution Type | Single Solution |
| SMILES | CN1CCN(CCCN2c3ccccc3Sc4ccccc24)CC1.OC(=O)\C=C/C(=O)O.OC(=O)\C=C/C(=O)O |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?