missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Perforin Monoclonal specifically detects Perforin in Human samples. It is validated for Immunohistochemistry (Paraffin).
Specifications
Specifications
| Antigen | Perforin |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | PRF1/7077R |
| Conjugate | DyLight 350 |
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Cytolysin, FLH2, HPLH2, HPLH2lymphocyte pore forming protein, Lymphocyte pore-forming protein, MGC65093, P1PFN1, perforin 1 (pore forming protein), perforin-1, PFPcytolysin |
| Host Species | Rabbit |
| Immunogen | Recombinant human Perforin protein fragment (around aa 413-552) (exact sequence is proprietary) (Uniprot: P14222) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?