missing translation for 'onlineSavingsMsg'
Learn More

Pericentrin Antibody, DyLight 755, R&D Systems™

Product Code. 18001157 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18001157 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18001157 Supplier Novus Biologicals Supplier No. NBP297596IR
  • ONLINE EXCLUSIVE PRICE
    494.16 €
    413.70 € / 0.1mL
    Save 80.46 €
    16% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

Pericentrin Polyclonal specifically detects Pericentrin in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pericentrin
Applications Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate DyLight 755
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Kendrin, KIAA0402, MOPD2, PCN, PCNT2pericentrin B, PCNTB, pericentrin, pericentrin 2 (kendrin), pericentrin, kendrin (KIAA0402)10KENPCTN2, pericentrin-2, pericentrin-380, pericentrin-B, SCKL4kendrin
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Pericentrin fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Centrosome Markers
Primary or Secondary Primary
Gene ID (Entrez) 5116
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.