missing translation for 'onlineSavingsMsg'
Learn More

Perilipin Antibody, PE, R&D Systems™

Product Code. 18606145 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
18606145 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18606145 Supplier Novus Biologicals Supplier No. NBP299070PE
  • ONLINE EXCLUSIVE PRICE
    463.47 €
    406.35 € / 0.1mL
    Save 57.12 €
    12% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

Perilipin Polyclonal specifically detects Perilipin in Human samples. It is validated for Immunohistochemistry/Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Formulation PBS
Host Species Rabbit
Gene ID (Entrez) 5346
Product Type Antibody
Form Purified
Product Line Novus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.