missing translation for 'onlineSavingsMsg'
Learn More

Peripheral Node Addressin Antibody (MECA-79R) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody has been used in 19 publications

Brand:  Novus Biologicals NBP2-78792SS

 View more versions of this product

Product Code. 18052626

  • ONLINE EXCLUSIVE PRICE
    261.00 €
    246.75 € / 0.10mg
    Save 14.25 €
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Peripheral Node Addressin Monoclonal specifically detects Peripheral Node Addressin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vivo assay.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Peripheral Node Addressin
Monoclonal
1 mg/mL
Western Blot 1:100-1:2000, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 31278331), Immunohistochemistry-Paraffin 1:100-1:500, Immunohistochemistry-Frozen 1:100-1:500, In vivo assay reported in scientific literature (PMID 30277476)
MECA-79, Peripheral Node Addressin, PNAd
Mouse lymph node stromal cells
0.1 mg
Primary
Store at -20°C.
IgG1
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In vitro Assay
MECA-79R
Unconjugated
PBS with 0.02% Sodium Azide
Rat
Protein A or G purified
Lipid and Metabolism
Human, Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.