missing translation for 'onlineSavingsMsg'
Learn More

Peripheral Node Addressin Antibody (MECA-79R) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody has been used in 19 publications

246.75 € - 551.25 €

Specifications

Antigen Peripheral Node Addressin
Clone MECA-79R
Concentration 1 mg/mL
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 31278331), Immunohistochemistry-Paraffin 1:100-1:500, Immunohistochemistry-Frozen 1:100-1:500, In vivo assay reported in scientific literature (PMID 30277476)
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In vitro Assay
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18052626
View Documents
Novus Biologicals
NBP2-78792SS
0.1 mg
ONLINE EXCLUSIVE PRICE
261.00 €
246.75 € / 0.10mg
Save 14.25 €
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18260068
View Documents
Novus Biologicals
NBP2-78792
0.5 mg
ONLINE EXCLUSIVE PRICE
583.00 €
551.25 € / 0.50mg
Save 31.75 €
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Peripheral Node Addressin Monoclonal specifically detects Peripheral Node Addressin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vivo assay.
Specifications

Specifications

Peripheral Node Addressin
1 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In vitro Assay
Unconjugated
Rat
Human, Mouse
MECA-79, Peripheral Node Addressin, PNAd
IgG1
Protein A or G purified
MECA-79R
Western Blot 1:100-1:2000, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 31278331), Immunohistochemistry-Paraffin 1:100-1:500, Immunohistochemistry-Frozen 1:100-1:500, In vivo assay reported in scientific literature (PMID 30277476)
Monoclonal
Purified
Lipid and Metabolism
PBS with 0.02% Sodium Azide
Mouse lymph node stromal cells
Primary
Store at -20°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.