missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peripherin Antibody (7C5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB300-220-0.025ML
This item is not returnable.
View return policy
Description
Peripherin Monoclonal specifically detects Peripherin in Mouse, Rat, Human (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Peripherin | |
| Monoclonal | |
| 1 mg/ml | |
| Western Blot 1:1000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:500 | |
| Neurofilament 4, neurofilament 4 (57kD), peripherin, PRPH1NEF4 | |
| Mouse | |
| 57 kDa | |
| 0.025 mL | |
| Primary | |
| Chicken. Expected to cross-react with all mammals due to sequence homology. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry | |
| 7C5 | |
| Unconjugated | |
| 50% PBS, 50% glycerol with 5mM Sodium Azide | |
| PRPH | |
| Recombinant rat Peripherin purified from E. coli. [UniProt# P21807] | |
| Tissue culture supernatant | |
| RUO | |
| 5630 | |
| Mammalia, Mouse, Chicken, Human (Negative), Rat | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction