missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peripherin Antibody (7C5), Novus Biologicals™
Mouse Monoclonal Antibody
156.00 € - 498.00 €
Specifications
| Antigen | Peripherin |
|---|---|
| Clone | 7C5 |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:500 |
| Applications | Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18222024
|
Novus Biologicals
NB300-220-0.025ML |
0.025 mL |
197.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18777863
|
Novus Biologicals
NB300-220 |
0.5 mL |
498.00 €
0.50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18519731
|
Novus Biologicals
NB300-220-0.1ml |
0.1 mL |
156.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Peripherin Monoclonal specifically detects Peripherin in Mouse, Rat, Human (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| Peripherin | |
| Western Blot 1:1000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:500 | |
| Monoclonal | |
| Supernatant | |
| RUO | |
| 50% PBS, 50% glycerol with 5mM Sodium Azide | |
| 5630 | |
| Recombinant rat Peripherin purified from E. coli. [UniProt# P21807] | |
| Tissue culture supernatant | |
| 57 kDa |
| 7C5 | |
| Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Mammalia, Mouse, Chicken, Human (Negative), Rat | |
| Neurofilament 4, neurofilament 4 (57kD), peripherin, PRPH1NEF4 | |
| PRPH | |
| Primary | |
| Chicken. Expected to cross-react with all mammals due to sequence homology. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title