missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PERK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-80930
This item is not returnable.
View return policy
Description
PERK Polyclonal antibody specifically detects PERK in Human, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| PERK | |
| Polyclonal | |
| Western Blot -Reported in scientific publication( PMID: 32587659), Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence -Reported in scientific publication (PMID: 23103912), Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Q9NZJ5, Q9NZJ5, Q9NZJ5, Q9NZJ5 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Chicken | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| DKFZp781H1925, EC 2.7.11.1, eukaryotic translation initiation factor 2 alpha kinase 3, eukaryotic translation initiation factor 2-alpha kinase 3, hsPEK, PEKPERKPancreatic eIF2-alpha kinase, PRKR-like endoplasmic reticulum kinase, WRS | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: NAWLEAPPEKWQEKMDEIWLKDESTDWPLSSPSPMDAPSVKIRRMDPFATKEHIEIIAPSPQRSRSFSVGISCDQTSSSESQFSPLEFSGMDHEDISESVDAAYNLQDSCLTDCDVEDGTMDGNDE | |
| 0.1 mL | |
| Protein Kinase | |
| 9451 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction