missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PERK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
369.00 € - 539.00 €
Specifications
| Antigen | PERK |
|---|---|
| Dilution | Western Blot -Reported in scientific publication( PMID: 32587659), Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence -Reported in scientific publication (PMID: 23103912), Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18408170
|
Novus Biologicals
NBP1-80930-25ul |
25 μL |
369.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18487420
|
Novus Biologicals
NBP1-80930 |
0.1 mL |
539.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PERK Polyclonal antibody specifically detects PERK in Human, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| PERK | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS (pH 7.2) and 40% Glycerol | |
| DKFZp781H1925, EC 2.7.11.1, eukaryotic translation initiation factor 2 alpha kinase 3, eukaryotic translation initiation factor 2-alpha kinase 3, hsPEK, PEKPERKPancreatic eIF2-alpha kinase, PRKR-like endoplasmic reticulum kinase, WRS | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: NAWLEAPPEKWQEKMDEIWLKDESTDWPLSSPSPMDAPSVKIRRMDPFATKEHIEIIAPSPQRSRSFSVGISCDQTSSSESQFSPLEFSGMDHEDISESVDAAYNLQDSCLTDCDVEDGTMDGNDE | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot -Reported in scientific publication( PMID: 32587659), Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence -Reported in scientific publication (PMID: 23103912), Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Chicken | |
| Q9NZJ5, Q9NZJ5, Q9NZJ5, Q9NZJ5 | |
| 9451 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title