missing translation for 'onlineSavingsMsg'
Learn More

Pethidine Acid Methyl Ester (Meperidine Acid Methyl Ester; Methyl 1-Methyl-4-phenylpiperidine-4-carboxylate) 0.1 mg/ml in Methanol, LoGiCal

Product Code. 30459248
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30459248

Brand: LoGiCal LGCAMP0315.0402

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Pethidine Acid Methyl Ester (Meperidine Acid Methyl Ester; Methyl 1-Methyl-4-phenylpiperidine-4-carboxylate) 0.1 mg/ml in Methanol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pethidine Acid Methyl Ester (Meperidine Acid Methyl Ester; Methyl 1-Methyl-4-phenylpiperidine-4-carboxylate)
CAS 28030-27-5
Analyte or Component Names Pethidine Acid Methyl Ester (Meperidine Acid Methyl Ester; Methyl 1-Methyl-4-phenylpiperidine-4-carboxylate)
Shipping Condition Room Temperature
Molecular Formula C14 H19 N O2
Certifications/Compliance ISO 17034
Synonym Methyl 1-Methyl-4-phenylpiperidine-4-carboxylate, Pethidine Hydrochloride Imp. D (EP), Pethidine Imp. D (EP), Pethidine Acid Methyl Ester, Meperidine Acid Methyl Ester, Methyl 1-Methyl-4-phenylpiperidine-4-carboxylate, Pethidine Hydrochloride Impurity D, Pethidine Impurity D
InChI Formula InChI=1 S/C14H19NO2/c1-15-10-8-14(9-11-15,13(16)17-2)12-6-4-3-5-7-12/h3-7 H,8-11H2,1-2H3
SMILES COC(=O)C1(CCN(C)CC1)c2ccccc2
IUPAC Name methyl 1-methyl-4-phenylpiperidine-4-carboxylate
Solvent or Matrix Methanol
Industry Type Forensics
Molecular Weight (g/mol) 233.31
Quantity 1 mL
Formula Weight 233.1416 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.