missing translation for 'onlineSavingsMsg'
Learn More

PGAM5 Antibody (CL0624) [DyLight 650], Novus Biologicals™

Product Code. 30297276 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30297276 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30297276 Supplier Novus Biologicals Supplier No. NBP252947C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PGAM5 Monoclonal antibody specifically detects PGAM5 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGAM5
Applications Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown
Classification Monoclonal
Clone CL0624
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias Bcl-XL-binding protein v68, BXLBv68, EC 3.1.3.16, MGC5352, phosphoglycerate mutase family member 5BXLBV68, serine/threonine-protein phosphatase PGAM5, mitochondrial
Host Species Mouse
Immunogen This antibody was developed using a recombinant protein derived from Q96HS1, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 192111
Target Species Human
Content And Storage Store at 4⪚C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.