missing translation for 'onlineSavingsMsg'
Learn More

PGC1 alpha/beta Antibody (SR1068), Novus Biologicals™

Product Code. 18613065 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18613065 50 μg 50µL
18633065 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18613065 Supplier Novus Biologicals Supplier No. NBP32170750ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

PGC1 alpha/beta Monoclonal antibody specifically detects PGC1 alpha/beta in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGC1 alpha/beta
Applications Western Blot
Classification Monoclonal
Clone SR1068
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias 4631412G21Rik;A830037N07Rik;ENSMUSG00000079510;ERRL1;Gm11133;LEM6;PERC;PGC1;PGC-1;PGC-1(alpha);PGC-1(beta);PGC1A;PGC-1alpha;PGC1B;PGC1beta;PGC-1v;Pgco1;PPARGC1;PPARGC1A;PPARGC1B
Host Species Rabbit
Immunogen A synthesized peptide derived from human PGC1 alpha/beta (Uniprot #: Q9UBK2, Q86YN6)
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.