missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PGC1 alpha/beta Monoclonal antibody specifically detects PGC1 alpha/beta in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | PGC1 alpha/beta |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | SR1068 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000 |
| Formulation | PBS, pH 7.4, 150mM NaCl, 50% glycerol. |
| Gene Alias | 4631412G21Rik;A830037N07Rik;ENSMUSG00000079510;ERRL1;Gm11133;LEM6;PERC;PGC1;PGC-1;PGC-1(alpha);PGC-1(beta);PGC1A;PGC-1alpha;PGC1B;PGC1beta;PGC-1v;Pgco1;PPARGC1;PPARGC1A;PPARGC1B |
| Host Species | Rabbit |
| Immunogen | A synthesized peptide derived from human PGC1 alpha/beta (Uniprot #: Q9UBK2, Q86YN6) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?