missing translation for 'onlineSavingsMsg'
Learn More

Phenacetin (p-Acetophenetidide), Lipomed

Product Code. 30457464
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457464 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457464 Supplier Lipomed Supplier No. LPMPHN978100
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Phenacetin (p-Acetophenetidide)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenacetin
CAS 62-44-2
Analyte or Component Names Phenacetin
Shipping Condition Room Temperature
Molecular Formula C10 H13 N O2
Synonym Phenacetin, p-Acetophenetidide, 4-(Acetylamino)phenetole, 4-Ethoxy-1-acetylaminobenzene, 4-Ethoxyacetanilide, 4'-Ethoxyacetanilide, 4'-Ethoxyacetanilide, 4'-Ethoxyacetanilide, 4'-Ethoxyacetanilide, 4'-Ethoxyacetanilide, 4'-Ethoxyacetanilide, 4'-Ethoxyacetanilide, 4'-Ethoxyacetanilide, 4'-Ethoxyacetanilide, 4'-Ethoxyacetanilide, 4'-Ethoxyacetanilide, 4'-Ethoxyacetanilide, 4'-Ethoxyacetanilide, 4'-Ethoxyacetanilide, 4'-Ethoxyacetanilide, 4'-Ethoxyacetanilide, 4'-Ethoxyacetanilide, 4'-Ethoxyacetanilide, 4'-Ethoxyacetanilide, 4'-Ethoxyacetanilide, 4'-Ethoxyacetanilide
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES CCOc1ccc(NC(=O)C)cc1
IUPAC Name N-(4-ethoxyphenyl)acetamide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 179.22
Quantity 100 mg
Formula Weight 179.0946 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.