missing translation for 'onlineSavingsMsg'
Learn More

Phenoxymethylpenicillin Potassium, Mikromol™

Product Code. 17972956
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 17972956

Brand: Mikromol MM0026.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Phenoxymethylpenicillin Potassium
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenoxymethylpenicillin Potassium
CAS 132-98-9
Analyte or Component Names Phenoxymethylpenicillin Potassium
Shipping Condition Ice pack (-20°C)
Molecular Formula C16 H17 N2 O5 S . K
Certifications/Compliance ISO 17034
Synonym Phenoxymethylpenicillin Potassium, (2S,5R,6R)-3,3-Dimethyl-7-oxo-6-[(2-phenoxyacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid potassium salt (1:1), 3,3-Dimethyl-7-oxo-6-(2-phenoxyacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid monopotassium salt, (2S,5R,6R)-3,3-Dimethyl-7-oxo-6-(2-phenoxyacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid monopotassium salt, Abbocillin VK, Acipen, Anapenil, Antibiocin, Apo-Pen VK, Apsin VK, Arcacil, Arcasin, Asillin, Beapen, Beepen Vk, Beromycin, Beromycin (penicillin), Beromycin 400, Betapen V, Betapen Vk, Calciopen K, Cilacil, Cilicaine VK, Cliacil, Compocillin VK, DQV-K, Distakaps V-K, Distaquaine V-K, Dowpen V-K, DuraPenicillin, Fenocin, Fenocin Forte, Fenospen, Fenoxcillin, Fenoxypen, Icipen, Isocillin, Ispenoral, Kavepenin, L.P.V., Ledercillin Vk, Len VK, Megacilina Oral, Megacillin-oral, Microcillin VK, Micropen VK, Milcopen, Monocillin E, Nadopen V, Newcillin, Novo VK, Novopen VK, Ocillin VK, Oracil VK, Oracillin VK, Orapen, Orvek, Ospen 250, Ospen KV, Ospeneff, P.V.O., PVK, Pedipen, Pen V-K, Pen-V, Pen-Vee K Powder, Pen-Vee-K, Pen-Vi-K, Penadur VK Mega, Penagen, Penapar VK, Pencompren, Penicillin V potassium, Penicillin V potassium salt, Penicillin VK, Penivoral, Penoxilin, Pentabs, Pentacillin, Pentranex, Penvikal, Penvisil, Pfizerpen Vk, Phenoxymethylpenicillin potassium, Phenoxymethylpenicillin potassium salt, Potassium penicillin V, Potassium penicillin V salt, Potassium phenoxymethylpenicillin, Potassium phenoxymethylpenicillinate, Primcillin, Qidpen VK, Rafapen V-K, Robicillin VK, Rocilin, Rocillin VK, Roscopenin, SK-Penicillin VK, Servipen V, Stabilin V-K, Stabillin VK syrup 125, Stabillin VK syrup 62.5, Sumapen VK, Suspen, Trepopen, Trepopen VK, Uticillin Vk, V-Cil K, V-Cillin K, V-Kal K, V-Pen, V-Penicillin Kalium, Vamosyn, Veetids, Vepen, Vepicombin, [2S-(2alpha,5alpha,6beta)]-3,3-Dimethyl-7-oxo-6-(2-phenoxyacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid monopotassium salt, Potassium 2,2-dimethyl-6alpha-phenoxyacetamidopenam-3alpha-carboxylate
InChI Formula InChI=1S/C16H18N2O5S.K/c1-16(2)12(15(21)22)18-13(20)11(14(18)24-16)17-10(19)8-23-9-6-4-3-5-7-9;/h3-7,11-12,14H,8H2,1-2H3,(H,17,19)(H,21,22);/q;+1/p-1/t11-,12+,14-;/m1./s1
SMILES [K+].CC1(C)S[C@@H]2[C@H](NC(=O)COc3ccccc3)C(=O)N2[C@H]1C(=O)[O-]
IUPAC Name potassium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenoxyacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular Weight (g/mol) 388.48
Quantity 250 mg
Formula Weight 388.0495 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.