missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenprobamate, Mikromol™
Description
Phenprobamate
Specifications
Specifications
| Chemical Name or Material | Phenprobamate |
| CAS | 673-31-4 |
| Analyte or Component Names | Phenprobamate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H13 N O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Benzenepropanol, 1-carbamate, 1-Propanol, 3-phenyl-, carbamate (8CI), Benzenepropanol, carbamate (9CI), Carbamic acid, 3-phenylpropyl ester (6CI,7CI), 3-Phenyl-1-propanol carbamate, 3-Phenylpropyl carbamate, Ansepron, Eirenal, Extacol, Gamaquil, MH 532, NSC 44682, NSC 50538, NSC 64270, Palmita, Phenprobamate, Proformiphen, Quamaquil, Spantol, Tranquil, gamma-Phenylpropyl carbamate |
| InChI Formula | InChI=1S/C10H13NO2/c11-10(12)13-8-4-7-9-5-2-1-3-6-9/h1-3,5-6H,4,7-8H2,(H2,11,12) |
| SMILES | NC(=O)OCCCc1ccccc1 |
| IUPAC Name | 3-phenylpropyl carbamate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?