missing translation for 'onlineSavingsMsg'
Learn More

Phentolamine Mesilate, LoGiCal

Product Code. 30460414
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30460414 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460414 Supplier LoGiCal Supplier No. LGCFOR1310.00

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phentolamine Mesilate
CAS 65-28-1
Analyte or Component Names Phentolamine Mesilate
Shipping Condition Room Temperature
Molecular Formula C17 H19 N3 O . C H4 O3 S
Certifications/Compliance ISO 9001
Synonym Phentolamine mesilate, 3-[[(4,5-Dihydro-1H-imidazol-2-yl)methyl](4-methylphenyl)amino]phenol methanesulfonate, Phenol, 3-[[(4,5-dihydro-1H-imidazol-2-yl)methyl](4-methylphenyl)amino]-, methanesulfonate (1:1), Phenol, 3-[[(4,5-dihydro-1H-imidazol-2-yl)methyl](4-methylphenyl)amino]-, monomethanesulfonate (salt) (9CI), Phenol, m-[N-(2-imidazolin-2-ylmethyl)-p-toluidino]-, monomethanesulfonate (salt) (8CI), Phentolamine mesylate, Phentolamine methanesulfonate, Phentolamine methanesulphonate, Regitin methanesulphonate, Regitine mesylate, Regitine methanesulfonate, Rogitine, Vasomax, m-[N-(2-Imidazolin-2-ylmethyl)-p-toluidino]phenol methanesulfonate
InChI Formula InChI=1S/C17H19N3O.CH4O3S/c1-13-5-7-14(8-6-13)20(12-17-18-9-10-19-17)15-3-2-4-16(21)11-15;1-5(2,3)4/h2-8,11,21H,9-10,12H2,1H3,(H,18,19);1H3,(H,2,3,4)
Solution Type Neat
SMILES Cc1ccc(cc1)N(CC2=NCCN2)c3cccc(O)c3.CS(=O)(=O)O
IUPAC Name 3-[N-(4,5-dihydro-1H-imidazol-2-ylmethyl)-4-methylanilino]phenol;methanesulfonic acid
Industry Type Clinical Testing
Molecular Weight (g/mol) 377.46
Quantity 10 mg
Formula Weight 377.1409 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.