missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Acetate, TRC

Product Code. 30006126
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006126 25 g 25g
30005259 50 g 50g
30012151 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006126 Supplier Toronto Research Chemicals Supplier No. TRCP31915025G
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-79-2
Molecular Formula C8 H8 O2
Molecular Weight (g/mol) 136.15
Synonym (Acetyloxy)benzene, Acetoxybenzene, NSC 27795, Phenol acetate, Phenyl acetate
IUPAC Name phenyl acetate
SMILES CC(=O)Oc1ccccc1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H8O2/c1-7(9)10-8-5-3-2-4-6-8/h2-6H,1H3
Formula Weight 136.0524
Chemical Name or Material Acetic acid-phenyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.