missing translation for 'onlineSavingsMsg'
Learn More

Phenyl phosphorodiamidate, 97%

Product Code. 11410064
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410064 10 g 10g
11420064 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410064 Supplier Thermo Scientific Alfa Aesar Supplier No. A13562.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7450-69-3 | C6H9N2O2P | 172.124 g/mol

Reagent for conversion of tautomeric keto-hydroxy groups (e.g. in N-heterocycles) directly to amino groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7450-69-3
Molecular Formula C6H9N2O2P
Molecular Weight (g/mol) 172.124
MDL Number MFCD00014767
InChI Key AYRRNFHDJUXLEQ-UHFFFAOYSA-N
Synonym phenyl phosphorodiamidate, phenylphosphorodiamidate, phenyl diamidophosphate, phosphorodiamidic acid, phenyl ester, diaminophosphoryl oxy benzene, phenylphosphorodiamidat, phosphoric phenyl ester diamide, phenyl phosphordiamidate, phenyl diamidophosphate #, diaminophenoxyphosphino-1-one
PubChem CID 81954
IUPAC Name diaminophosphoryloxybenzene
SMILES C1=CC=C(C=C1)OP(=O)(N)N

Specifications

Melting Point 180°C to 185°C
Quantity 10 g
Beilstein 608629
Sensitivity Air and light sensitive
Formula Weight 172.12
Percent Purity ≥98%
Chemical Name or Material Phenyl phosphorodiamidate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.