missing translation for 'onlineSavingsMsg'
Learn More

Phenylalanine (L-Phenylalanine), Mikromol™

Product Code. 17938764
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17938764 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17938764 Supplier Mikromol Supplier No. MM1560.00
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Phenylalanine (L-Phenylalanine)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Phenylalanine
CAS 63-91-2
Analyte or Component Names L-Phenylalanine
Shipping Condition Room Temperature
Molecular Formula C9 H11 N O2
Certifications/Compliance ISO 17034
Synonym Nateglinide Imp. D (EP), Tyrosine Imp. A (EP), L-Phenylalanine, Alanine, phenyl-, L- (4CI,7CI,8CI), (-)-Phenylalanine, (2S)-2-Amino-3-phenylpropanoic acid, (S)-(-)-Phenylalanine, (S)-2-Amino-3-phenylpropanoic acid, (S)-2-Amino-3-phenylpropionic acid, (S)-Phenylalanine, (S)-alpha-Amino-beta-phenylpropionic acid, (S)-alpha-Aminobenzenepropanoic acid, (S)-alpha-Aminohydrocinnamic acid, 11: PN: US20090069547 PAGE: 10 claimed protein, 151: PN: US20070015162 SEQID: 161 claimed protein, 175: PN: EP2071334 SEQID: 191 claimed protein, 175: PN: WO2009077864 SEQID: 191 claimed protein, 3-Phenyl-L-alanine, 3-Phenylalanine, 34: PN: AU2005201745 SEQID: 22 claimed protein, 356: PN: US20070026454 SEQID: 366 claimed protein, 49: PN: WO2010069074 SEQID: 47 claimed sequence, Antibiotic FN 1636, Benzenepropanoic acid, alpha-amino-, (S)-, L-(-)-Phenylalanine, L-Alanine, 3-phenyl-, NSC 79477, Phenyl-alpha-alanine, Phenylalanine, beta-Phenyl-L-alanine, beta-Phenyl-alpha-alanine, Aspartame Imp. C (EP), GV 112054X, Aspartame Impurity C (EP), Nateglinide Impurity D, Tyrosine Impurity A
InChI Formula InChI=1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)/t8-/m0/s1
SMILES N[C@@H](Cc1ccccc1)C(=O)O
IUPAC Name (2S)-2-amino-3-phenylpropanoic acid
Molecular Weight (g/mol) 165.19
Quantity 250 mg
Formula Weight 165.079 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.