missing translation for 'onlineSavingsMsg'
Learn More

Phenylalanine, Mikromol™

Product Code. 30050163
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050163 25 mg 25mg
17978914 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050163 Supplier Mikromol Supplier No. MM0314.030025
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Phenylalanine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DL-Phenylalanine
CAS 150-30-1
Analyte or Component Names DL-Phenylalanine
Shipping Condition Room Temperature
Molecular Formula C9 H11 N O2
Certifications/Compliance ISO 17025
Synonym Phenylalanine, Aspartame Imp. C (EP) as Racemate, Alanine, phenyl-, DL- (8CI), 2-Amino-3-phenylpropanoic acid, DL-2-Amino-3-phenylpropanoic acid, DL-2-Amino-3-phenylpropionic acid, DL-3-Phenylalanine, DL-Phenylalanine, DL-beta-Phenyl-alpha-alanine, DLPA, NSC 9959, Alanine, beta-phenyl- (3CI), (+/-)-3-Phenyl-2-aminopropanoic acid, (+/-)-Phenylalanine, (+/-)-beta-Phenyl-alpha-alanine, DL-alpha-Amino-beta-phenylpropionic acid, Aspartame Impurity C as racemate
InChI Formula InChI=1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)
SMILES NC(Cc1ccccc1)C(=O)O
IUPAC Name 2-amino-3-phenylpropanoic acid
Molecular Weight (g/mol) 165.19
Quantity 25 mg
Formula Weight 165.079 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.