missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98955-100ul
This item is not returnable.
View return policy
Description
PHF1 Polyclonal antibody specifically detects PHF1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| PHF1 | |
| Polyclonal | |
| Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:1500-1:50000 | |
| hPCl1, MTF2L2, PCL1, PHD finger protein 1, PHF2, Polycomb-like protein 1, Protein PHF1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PHF1. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 5252 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction