missing translation for 'onlineSavingsMsg'
Learn More

PHF1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

227.00 € - 326.00 €

Specifications

Antigen PHF1
Dilution Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:1500-1:50000
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18647605
View Documents
Novus Biologicals
NBP2-98955-100ul
100 μg
326.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18682676
View Documents
Novus Biologicals
NBP2-98955-50ul
50 μg
227.00 €
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PHF1 Polyclonal antibody specifically detects PHF1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

PHF1
Western Blot, Immunofluorescence
Unconjugated
Rabbit
Human
hPCl1, MTF2L2, PCL1, PHD finger protein 1, PHF2, Polycomb-like protein 1, Protein PHF1
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PHF1.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:1500-1:50000
Polyclonal
Purified
RUO
0.2 μm filtered solution in PBS
5252
IgG
Antigen and protein A Affinity-purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.