missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PHF22 Recombinant Rabbit Monoclonal Antibody (2D3T4)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557955

 View more versions of this product

Product Code. 30284061

  • 176.00 € / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MAATVNLELD PIFLKALGFL HSKSKDSAEK LKALLDESLA RGIDSSYRPS QKDVEPPKIS STKNISIKQE PKISSSLPSG NNNGKVLTTE KVKKEAEKRP ADKMKSDITE GVDIPKKPRL EKPETQSSPI TVQSSKDLPM ADLSSFEETS ADDFAMEMGL ACVVCRQMMV ASGNQLVECQ ECHNLYHRDC HKPQVTDKEA NDPRLVWYCA RCTRQMKRMA QKTQKPPQKP.

INTS12 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit and mediates 3-prime end processing of small nuclear RNAs U1 and U2.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PHF22
Recombinant Monoclonal
0.58 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Q9D168
Ints12
Recombinant fusion protein containing a sequence corresponding to amino acids 1-230 of human INTS12 (NP_065128.2).
20 μL
Primary
Mouse
Antibody
IgG
ELISA, Western Blot
2D3T4
Unconjugated
Ints12
1110020M19Rik; 2810027J24Rik; 4930529N21Rik; A230056J18Rik; AI666778; hypothetical nuclear factor SBBI22; hypothetical protein LOC415251; int12; Integrator complex subunit 12; INTS12; PHD finger protein 22; PHF22; SBBI22; Unknown (protein for MGC:134302); wu:fi17f05; zgc:86602
Rabbit
Protein A
RUO
71793
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.