Learn More
Invitrogen™ Phospho-Cyclin B1 (Ser126) Recombinant Rabbit Monoclonal Antibody (24GB4310)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557572
 View more versions of this product
Product Code. 30275181
  • 564.00 € / 100µL
Description
Phospho-Cyclin B1 (Ser126) Recombinant Monoclonal Antibody for Western Blot, ICC/IF
Cyclins bind to and regulate the activity of the Cyclin dependent protein kinases (CDKs). Cyclin B1 or CCNB1 is a regulatory protein involved in mitosis. It is essential for the control of the cell cycle at the G2/M transition. Cyclin B1 complexes with p34 (cdc2) to form the maturation-promoting factor (MPF). Two alternative transcripts have been found, a constitutively expressed transcript and a cell cycle-regulated transcript, that is expressed predominantly during G2/M phase. Cyclin B1 is not ubiquitinated during G2/M phase, resulting in its steady accumulation during G2 phase, followed by abrupt APC dependent destruction at the end of mitosis. Destruction of Cyclin B1 is required for cell cycle progression. Cyclin B1 is overexpressed in various cancers, including breast, prostate, and non-small cell lung cancer.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Phospho-Cyclin B1 (Ser126)
Recombinant Monoclonal
Unconjugated
Ccnb1
Ccn-2; CCNB; Ccnb1; Ccnb1-rs1; Ccnb1-rs13; Cycb; Cycb1; Cycb1-rs1; Cycb-4; Cycb-5; cyclin B; cyclin B1; cyclin B1, related sequence 1; cyclin B1, related sequence 13; G2/mitotic-specific cyclin B1; G2/mitotic-specific cyclin-B1; I79_013589; OTTHUMP00000221981; OTTHUMP00000223023
Rabbit
Affinity chromatography
RUO
891
-20°C
Liquid
Western Blot, Immunocytochemistry
24GB4310
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P14635
Ccnb1
A synthesized peptide derived from human Phospho-Cyclin B1 (Ser126) (aa 120-133).