missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-EIF2S1 (Ser51) Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28740-1-AP-100UL
This item is not returnable.
View return policy
Descripción
The Phospho-EIF2S1 (Ser51) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-EIF2S1 (Ser51). This antibody recognizes human, mouse, rat antigen. The Phospho-EIF2S1 (Ser51) antibody has been validated for the following applications: WB, ELISA analysis.Especificaciones
| EIF2S1 (Ser51) | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 1965 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, ELISA | |
| 570 μg/mL | |
| EIF-2; EIF-2A; EIF-2alpha; EIF2; EIF2A | |
| Peptide | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 36 kDa | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur