missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-EIF2S1 (Ser51) Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
135.00 € - 360.00 €
Specifications
| Antigen | EIF2S1 (Ser51) |
|---|---|
| Concentration | 570 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30237955
|
Proteintech
28740-1-AP-20UL |
20 μL |
135.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30237915
|
Proteintech
28740-1-AP-100UL |
100 μL |
360.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Phospho-EIF2S1 (Ser51) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-EIF2S1 (Ser51). This antibody recognizes human, mouse, rat antigen. The Phospho-EIF2S1 (Ser51) antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| EIF2S1 (Ser51) | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| EIF-2; EIF-2A; EIF-2alpha; EIF2; EIF2A | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
| 570 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 1965 | |
| Peptide | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title