missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-eIF4B (Ser406) Recombinant Rabbit Monoclonal Antibody (23GB3905), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553663
This item is not returnable.
View return policy
Description
The eukaryotic translation initiation factor 4B (eIF4B) plays a critical role in recruiting the 40S ribosomal subunit to the mRNA. It functions in close association with eIF4F and eIF4A. It binds near the 5'-terminal cap of mRNA in the presence of eIF4F and ATP. It promotes the ATPase activity and the ATP-dependent RNA unwinding activity of both eIF4A and eIF4F.
Specifications
| Phospho-eIF4B (Ser406) | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Eif4b | |
| 2310046H11Rik; AL024095; C85189; Eif4a2; EIF4B; eIF-4B; Eukaryotic translation initiation factor 4B; hCG_2014609; hypothetical protein; PRO1843 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1975, 300253, 75705 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3905 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P23588, Q8BGD9 | |
| Eif4b | |
| A synthetic peptide of human Phospho-eIF4B (Ser406) (399-413AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion