missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-eIF4B (Ser406) Recombinant Rabbit Monoclonal Antibody (23GB3905), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
207.00 € - 562.00 €
Specifications
| Antigen | Phospho-eIF4B (Ser406) |
|---|---|
| Clone | 23GB3905 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
30089959
|
Invitrogen™
MA553662 |
100 μL |
562.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087711
|
Invitrogen™
MA553663 |
20 μL |
207.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
Phospho-eIF4B (Ser406) Recombinant Monoclonal antibody specifically detects Phospho-eIF4B (Ser406) in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpezifikation
| Phospho-eIF4B (Ser406) | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P23588, Q8BGD9 | |
| 1975, 300253, 75705 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3905 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 2310046H11Rik; AL024095; C85189; Eif4a2; EIF4B; eIF-4B; Eukaryotic translation initiation factor 4B; hCG_2014609; hypothetical protein; PRO1843 | |
| A synthetic peptide of human Phospho-eIF4B (Ser406) (399-413AA). | |
| Primary | |
| -20°C | |
| Eif4b |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts