Learn More
Phospho-TDP43 (Ser409/410) Rabbit anti-Human, Unconjugated, Clone: 6M10, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 80007-1-RR-100UL
Description
The Phospho-TDP43 (Ser409/410) antibody from Proteintech is a rabbit recombinant antibody generated with peptide of human Phospho-TDP43 (Ser409/410). This antibody recognizes human, mouse antigen. The Phospho-TDP43 (Ser409/410) antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| Phospho-TDP43 (Ser409/410) | |
| Monoclonal | |
| Unconjugated | |
| 100 μL | |
| 23435 | |
| IgG |
| Western Blot, Immunohistochemistry | |
| 6M10 | |
| Rabbit | |
| Primary | |
| Human, Mouse |