Learn More
Phospho-TDP43 (Ser409/410) Rabbit anti-Human, Unconjugated, Clone: 6M10, Proteintech
Rabbit Recombinant Antibody
149.00 € - 439.00 €
Specifications
| Antigen | Phospho-TDP43 (Ser409/410) |
|---|---|
| Clone | 6M10 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16800436
|
Proteintech
80007-1-RR-20UL |
20 μL |
149.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16890426
|
Proteintech
80007-1-RR-100UL |
100 μL |
439.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Phospho-TDP43 (Ser409/410) antibody from Proteintech is a rabbit recombinant antibody generated with peptide of human Phospho-TDP43 (Ser409/410). This antibody recognizes human, mouse antigen. The Phospho-TDP43 (Ser409/410) antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| Phospho-TDP43 (Ser409/410) | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Human, Mouse | |
| IgG |
| 6M10 | |
| Monoclonal | |
| Rabbit | |
| 23435 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.